4-Brom-m-toluylsäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 4-Bromo-3-methylbenzoic acid, 2-Bromo-5-carboxytoluene, 4-Bromo-m-toluic acid

AS1497G100 AS1497G5
ACTIAS1497G100EA 0 CHF
ACTIAS1497G100 ACTIAS1497G5
4-Brom-m-toluylsäure
4-Brom-m-toluylsäure
Formel: C₈H₇BrO₂
MW: 215,05 g/mol
MDL Number: MFCD00039526
CAS-Nummer: 7697-28-1
EINECS: 231-713-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR