2-Chlor-4-pyridinboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloropyridine-4-boronic acid pinacol ester, 2-(2-Chloro-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Chloropyridine-4-boronic acid Pinacol ester

AS33968G1 AS33968G10 AS33968G5
ACTIAS33968G1EA 0 CHF
ACTIAS33968G1 ACTIAS33968G10 ACTIAS33968G5
2-Chlor-4-pyridinboronsäurepinakolester
2-Chlor-4-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BClNO₂
MW: 239,51 g/mol
MDL Number: MFCD04039870
CAS-Nummer: 458532-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR