6-Brom-2-pyridinboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Bromo-2-pyridylboronic acid Pinacol ester, 6-Bromopyridine-2-boronic acid pinacol ester, 2-(6-Bromo-2-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

AS31334G1 AS31334G5
ACTIAS31334G1EA 0 CHF
ACTIAS31334G1 ACTIAS31334G5
6-Brom-2-pyridinboronsäurepinakolester
6-Brom-2-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BBrNO₂
MW: 283.96 g/mol
MDL Number: MFCD06796177
CAS-Nummer: 651358-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR