3-Pyridinboronsäurepinakolester
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Pyridine-3-boronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
AS21967G25
AS21967G5
ACTIAS21967G25EA
68
CHF
ACTIAS21967G25
ACTIAS21967G5
3-Pyridinboronsäurepinakolester
3-Pyridinboronsäurepinakolester
Formel:
C₁₁H₁₆BNO₂ MW: 205,06 g/mol |
MDL Number:
MFCD03084758 CAS-Nummer: 329214-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...