2-Fluor-5-nitrophenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-5-nitrobenzeneboronic acid

AS51135G1 AS51135G5
ACTIAS51135G1EA 0 CHF
ACTIAS51135G1 ACTIAS51135G5
2-Fluor-5-nitrophenylboronsäure
2-Fluor-5-nitrophenylboronsäure
Formel: C₆H₅BFNO₄
MW: 184,92 g/mol
MDL Number: MFCD03092929
CAS-Nummer: 819849-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR