2-Chlor-5-nitrophenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS35085G1 AS35085G10 AS35085G5
ACTIAS35085G1EA 0 CHF
ACTIAS35085G1 ACTIAS35085G10 ACTIAS35085G5
2-Chlor-5-nitrophenylboronsäure
2-Chlor-5-nitrophenylboronsäure
Formel: C₆H₅BClNO₄
MDL Number: MFCD03095323
CAS-Nummer: 867333-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR