2-Chlor-5-fluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-5-fluorobenzeneboronic acid

AS35444G1 AS35444G5
ACTIAS35444G1EA 0 CHF
ACTIAS35444G1 ACTIAS35444G5
2-Chlor-5-fluorphenylboronsäure
2-Chlor-5-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
MW: 174,37 g/mol
MDL Number: MFCD06656272
CAS-Nummer: 444666-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR