(2-Fluor-4-methoxyphenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-4-methoxyphenylacetic acid

AS40239G1 AS40239G5
ACTIAS40239G1EA 0 CHF
ACTIAS40239G1 ACTIAS40239G5
(2-Fluor-4-methoxyphenyl)essigsäure
(2-Fluor-4-methoxyphenyl)essigsäure
Formel: C₉H₉FO₃
MW: 184,17 g/mol
MDL Number: MFCD06800744
CAS-Nummer: 883531-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR