2-Fluor-3-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-3-methoxybenzeneboronic acid, (2-Fluoro-3-methoxyphenyl)boronic acid

AS51368G1 AS51368G100 AS51368G5
ACTIAS51368G1EA 0 CHF
ACTIAS51368G1 ACTIAS51368G100 ACTIAS51368G5
2-Fluor-3-methoxyphenylboronsäure
2-Fluor-3-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
MW: 169,95 g/mol
MDL Number: MFCD02094728
CAS-Nummer: 352303-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR