2,6-Difluor-4-methoxyphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
AS34059G1
AS34059G5
ACTIAS34059G1EA
0
CHF
ACTIAS34059G1
ACTIAS34059G5
2,6-Difluor-4-methoxyphenylboronsäure
2,6-Difluor-4-methoxyphenylboronsäure
Formel:
C₇H₇BF₂O₃ |
MDL Number:
MFCD05664299 CAS-Nummer: 406482-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...