3-Phenylbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Biphenyl-3-carboxylic acid, 3-Biphenylcarboxylic acid, 1,1'-Biphenyl-3-carboxylic acid, [1,1'-Biphenyl]-3-carboxylic acid

AS33134G1 AS33134G5
ACTIAS33134G1EA 0 CHF
ACTIAS33134G1 ACTIAS33134G5
3-Phenylbenzoesäure
3-Phenylbenzoic acid
Formel: C₁₃H₁₀O₂
MW: 198,22 g/mol
MDL Number: MFCD00045846
CAS-Nummer: 716-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR