2-Chlorphenylboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chlorophenylboronic acid Pinacol ester, 2-(2-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

AS22753G1 AS22753G25 AS22753G5
ACTIAS22753G1EA 18 CHF
ACTIAS22753G1 ACTIAS22753G25 ACTIAS22753G5
2-Chlorphenylboronsäurepinakolester
2-Chlorphenylboronsäurepinakolester
Formel: C₁₂H₁₆BClO₂
MW: 238.52 g/mol
MDL Number: MFCD05663846
CAS-Nummer: 870195-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR