3-Iodbenzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
AS23309G25
AS23309G5
ACTIAS23309G25EA
0
CHF
ACTIAS23309G25
ACTIAS23309G5
3-Iodbenzaldehyd
3-Iodobenzaldehyde
Formel:
C₇H₅IO |
MDL Number:
MFCD00039573 CAS-Nummer: 696-41-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...