4-Brom-2-(trifluormethyl)benzonitril
Lieferant: ACTIVATE SCIENTIFIC
AS32298G1
AS32298G5
ACTIAS32298G1EA
0
CHF
ACTIAS32298G1
ACTIAS32298G5
4-Brom-2-(trifluormethyl)benzonitril
4-Brom-2-(trifluormethyl)benzonitril
Formel:
C₈H₃BrF₃N |
MDL Number:
MFCD06659466 CAS-Nummer: 191165-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...