2-Brom-4-methylbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS33897G1 AS33897G10 AS33897G5
ACTIAS33897G1EA 0 CHF
ACTIAS33897G1 ACTIAS33897G10 ACTIAS33897G5
2-Brom-4-methylbenzaldehyd
2-Bromo-4-methylbenzaldehyde
Formel: C₈H₇BrO
MW: 199,05 g/mol
MDL Number: MFCD04039890
CAS-Nummer: 824-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR