2-(Trimethylsilyl)thiazol

Lieferant: ACTIVATE SCIENTIFIC

AS50278G1 AS50278G5
ACTIAS50278G1EA 0 CHF
ACTIAS50278G1 ACTIAS50278G5
2-(Trimethylsilyl)thiazol
2-(Trimethylsilyl)thiazole
Formel: C₆H₁₁NSSi
MW: 157,31 g/mol
MDL Number: MFCD00066274
CAS-Nummer: 79265-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR