2-Brom-5-thiazolcarbonsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Bromothiazole-5-carboxylic acid
AS50074G1
AS50074G5
ACTIAS50074G1EA
0
CHF
ACTIAS50074G1
ACTIAS50074G5
2-Brom-5-thiazolcarbonsäure
2-Bromo-5-thiazolecarboxylic acid
Formel:
C₄H₂BrNO₂S MW: 208,04 g/mol |
MDL Number:
MFCD04115730 CAS-Nummer: 54045-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...