2-Brom-4-thiazolcarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-1,3-thiazole-4-carboxylic acid, 2-Bromothiazole-4-carboxylic acid

AS50073G1 AS50073G5
ACTIAS50073G1EA 0 CHF
ACTIAS50073G1 ACTIAS50073G5
2-Brom-4-thiazolcarbonsäure
2-Bromo-4-thiazolecarboxylic acid
Formel: C₄H₂BrNO₂S
MW: 208,04 g/mol
MDL Number: MFCD04115729
CAS-Nummer: 5198-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR