2-Brom-6-fluorbenzothiazol

Lieferant: ACTIVATE SCIENTIFIC

AS31117G1 AS31117G5
ACTIAS31117G1EA 0 CHF
ACTIAS31117G1 ACTIAS31117G5
2-Brom-6-fluorbenzothiazol
2-Bromo-6-fluorobenzothiazole
Formel: C₇H₃BrFNS
MDL Number: MFCD07783786
CAS-Nummer: 152937-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR