1-Boc-5-aminoindol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Amino-1H-indole, N1-BOC protected, 1-Boc-5-Aminoindole
AS30520G1
AS30520G5
ACTIAS30520G1EA
0
CHF
ACTIAS30520G1
ACTIAS30520G5
1-Boc-5-aminoindol
1-Boc-5-aminoindol
Formel:
C₁₃H₁₆N₂O₂ MW: 232,28 g/mol |
MDL Number:
MFCD04114746 CAS-Nummer: 166104-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...