6-(Trifluormethyl)oxindol

Lieferant: ACTIVATE SCIENTIFIC

AS32287G1 AS32287G5
ACTIAS32287G1EA 0 CHF
ACTIAS32287G1 ACTIAS32287G5
6-(Trifluormethyl)oxindol
6-(Trifluormethyl)oxindol
Formel: C₉H₆F₃NO
MDL Number: MFCD01075220
CAS-Nummer: 1735-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR