(R)-(+)-4-Brom-α-methylbenzylalkohol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (R)-1-(4-Bromophenyl)ethanol

AS36803G1 AS36803G5
ACTIAS36803G1EA 0 CHF
ACTIAS36803G1 ACTIAS36803G5
(R)-(+)-4-Brom-α-methylbenzylalkohol
(R)-(+)-4-Brom-α-methylbenzylalkohol
Formel: C₈H₉BrO
MDL Number: MFCD06201861
CAS-Nummer: 76155-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR