(R)-(+)-2-Brom-α-methylbenzylalkohol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (R)-(+)-1-(2-Bromophenyl)ethanol, (R)-2-Bromo-alpha-methylbenzyl Alcohol

AS36205G1 AS36205G5
ACTIAS36205G1EA 0 CHF
ACTIAS36205G1 ACTIAS36205G5
(R)-(+)-2-Brom-α-methylbenzylalkohol
(R)-(+)-2-Brom-α-methylbenzylalkohol
Formel: C₈H₉BrO
MDL Number: MFCD00216591
CAS-Nummer: 76116-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR