1-Boc-2-piperidon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: N-Boc-2-Piperidone, 1-N-Boc-2-piperidone

AS32558G1 AS32558G10 AS32558G5
ACTIAS32558G1EA 0 CHF
ACTIAS32558G1 ACTIAS32558G10 ACTIAS32558G5
1-Boc-2-piperidon
1-Boc-2-piperidon
Formel: C₁₀H₁₇NO₃
MW: 199.25 g/mol
MDL Number: MFCD02179046
CAS-Nummer: 85908-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR