4-Brom-3-nitrobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS31518G1 AS31518G10 AS31518G5
ACTIAS31518G1EA 0 CHF
ACTIAS31518G1 ACTIAS31518G10 ACTIAS31518G5
4-Brom-3-nitrobenzaldehyd
4-Bromo-3-nitrobenzaldehyde
Formel: C₇H₄BrNO₃
MW: 230,02 g/mol
MDL Number: MFCD00204031
CAS-Nummer: 163596-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR