3-Brom-5-nitrobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS30847G1 AS30847G5
ACTIAS30847G1EA 0 CHF
ACTIAS30847G1 ACTIAS30847G5
3-Brom-5-nitrobenzaldehyd
3-Bromo-5-nitrobenzaldehyde
Formel: C₇H₄BrNO₃
MW: 230,02 g/mol
MDL Number: MFCD00100097
CAS-Nummer: 355134-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR