Diethylfluormalonat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
1,3-diethyl 2-fluoropropanedioate
AS33970G1
AS33970G5
ACTIAS33970G1EA
0
CHF
ACTIAS33970G1
ACTIAS33970G5
Diethylfluormalonat
Diethylfluormalonat
Formel:
C₇H₁₁FO₄ |
MDL Number:
MFCD00009139 CAS-Nummer: 685-88-1 EINECS: 211-684-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...