3-Nitroanthranilsäure

Lieferant: ACTIVATE SCIENTIFIC

AS22111G100 AS22111G25
ACTIAS22111G100EA 161 CHF
ACTIAS22111G100 ACTIAS22111G25
3-Nitroanthranilsäure
3-Nitroanthranilic acid
Formel: C₇H₆N₂O₄
MW: 182,14 g/mol
MDL Number: MFCD00024261
CAS-Nummer: 606-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR