2-Brom-6-nitroanilin

Lieferant: ACTIVATE SCIENTIFIC

AS35634G1 AS35634G5
ACTIAS35634G1EA 0 CHF
ACTIAS35634G1 ACTIAS35634G5
2-Brom-6-nitroanilin
2-Brom-6-nitroanilin
Formel: C₆H₅BrN₂O₂
MDL Number: MFCD09263476
CAS-Nummer: 59255-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR