3-Brom-2-fluoranilin

Lieferant: ACTIVATE SCIENTIFIC

AS32421G1 AS32421G5
ACTIAS32421G1EA 0 CHF
ACTIAS32421G1 ACTIAS32421G5
3-Brom-2-fluoranilin
3-Brom-2-fluoranilin
Formel: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD09864700
CAS-Nummer: 58534-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR