4-Benzyloxyindol
Lieferant: ACTIVATE SCIENTIFIC
AS50060G100
AS50060G5
ACTIAS50060G100EA
0
CHF
ACTIAS50060G100
ACTIAS50060G5
4-Benzyloxyindol
4-Benzyloxyindol
Formel:
C₁₅H₁₃NO MW: 223,27 g/mol |
MDL Number:
MFCD00047200 CAS-Nummer: 20289-26-3 EINECS: 243-690-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...