6-Fluorsalicylaldehyd
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Fluoro-6-hydroxybenzaldehyde
AS32361G1
AS32361G5
ACTIAS32361G1EA
0
CHF
ACTIAS32361G1
ACTIAS32361G5
6-Fluorsalicylaldehyd
6-Fluorsalicylaldehyd
Formel:
C₇H₅FO₂ |
MDL Number:
MFCD01090996 CAS-Nummer: 38226-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...