5-Brombenzothiazol
Lieferant: ACTIVATE SCIENTIFIC
AS31968G1
AS31968G5
ACTIAS31968G1EA
0
CHF
ACTIAS31968G1
ACTIAS31968G5
5-Brombenzothiazol
5-Bromobenzothiazole
Formel:
C₇H₄BrNS |
MDL Number:
MFCD03001442 CAS-Nummer: 768-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...