2-Amino-4-(2-methyl-2-propanyl)phenol

Lieferant: ACTIVATE SCIENTIFIC

AS25473G100 AS25473G25
ACTIAS25473G100EA 0 CHF
ACTIAS25473G100 ACTIAS25473G25
2-Amino-4-(2-methyl-2-propanyl)phenol
2-Amino-4-tert-butylphenol
Formel: C₁₀H₁₅NO
MW: 165,24 g/mol
MDL Number: MFCD00007698
CAS-Nummer: 1199-46-8
EINECS: 214-844-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR