4,6-Dimethoxyindol

Lieferant: ACTIVATE SCIENTIFIC

AS36123G1 AS36123G5
ACTIAS36123G1EA 0 CHF
ACTIAS36123G1 ACTIAS36123G5
4,6-Dimethoxyindol
4,6-Dimethoxyindole
Formel: C₁₀H₁₁NO₂
MDL Number: MFCD00128213
CAS-Nummer: 23659-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR