4-Amino-2-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS30586G1 AS30586G5
ACTIAS30586G1EA 0 CHF
ACTIAS30586G1 ACTIAS30586G5
4-Amino-2-fluorpyridin
4-Amino-2-fluoropyridine
Formel: C₅H₅FN₂
MW: 112,11 g/mol
MDL Number: MFCD03095284
CAS-Nummer: 18614-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR