4,6-Difluorindol

Lieferant: ACTIVATE SCIENTIFIC

AS35177G1 AS35177G5
ACTIAS35177G1EA 0 CHF
ACTIAS35177G1 ACTIAS35177G5
4,6-Difluorindol
4,6-Difluoroindole
Formel: C₈H₅F₂N
MDL Number: MFCD01075213
CAS-Nummer: 199526-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR