3-Hydroxy-4-iodbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS34191G1 AS34191G5
ACTIAS34191G1EA 0 CHF
ACTIAS34191G1 ACTIAS34191G5
3-Hydroxy-4-iodbenzoesäure
3-Hydroxy-4-iodbenzoesäure
Formel: C₇H₅IO₃
MW: 264,02 g/mol
MDL Number: MFCD02068386
CAS-Nummer: 58123-77-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR