3-Iodbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS24782G100 AS24782G25
ACTIAS24782G100EA 249 CHF
ACTIAS24782G100 ACTIAS24782G25
3-Iodbenzonitril
3-Iodbenzonitril
Formel: C₇H₄IN
MW: 229,02 g/mol
MDL Number: MFCD00079762
CAS-Nummer: 69113-59-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR