2-Fluor-4-nitroanilin
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS24997G100
AS24997G25
ACTIAS24997G100EA
100
CHF
ACTIAS24997G100
ACTIAS24997G25
2-Fluor-4-nitroanilin
2-Fluor-4-nitroanilin
Formel:
C₆H₅FN₂O₂ MW: 156,12 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00034560 CAS-Nummer: 369-35-7 EINECS: 206-719-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...