4-Fluor-2-nitrobenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS23396G100 AS23396G25
ACTIAS23396G100EA 0 CHF
ACTIAS23396G100 ACTIAS23396G25
4-Fluor-2-nitrobenzoesäure
4-Fluoro-2-nitrobenzoic acid
Formel: C₇H₄FNO₄
MDL Number: MFCD00024300
CAS-Nummer: 394-01-4
EINECS: 206-890-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR