2,4,6-Trifluorbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS24177G100 AS24177G25
ACTIAS24177G100EA 0 CHF
ACTIAS24177G100 ACTIAS24177G25
2,4,6-Trifluorbenzoesäure
2,4,6-Trifluorobenzoic acid
Formel: C₇H₃F₃O₂
MW: 176,09 g/mol
Siedepunkt: 237 °C (1013 hPa)
Melting Pt: 144…145 °C
MDL Number: MFCD00042398
CAS-Nummer: 28314-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR