3,4,5-Trifluorbenzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
AS35015G1
AS35015G10
AS35015G5
ACTIAS35015G1EA
0
CHF
ACTIAS35015G1
ACTIAS35015G10
ACTIAS35015G5
3,4,5-Trifluorbenzaldehyd
3,4,5-Trifluorobenzaldehyde
Formel:
C₇H₃F₃O MW: 160,1 g/mol Siedepunkt: 176…183 °C (1013 hPa) Melting Pt: 25 °C Dichte: 1.42 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00083523 CAS-Nummer: 132123-54-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...