1,3-Dichlor-5-iodbenzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3,5-Dichloroiodobenzene

AS21043G100 AS21043G25
ACTIAS21043G100EA 123 CHF
ACTIAS21043G100 ACTIAS21043G25
1,3-Dichlor-5-iodbenzol
1,3-Dichlor-5-iodbenzol
Formel: C₆H₃Cl₂I
MW: 272,9 g/mol
Siedepunkt: 127 °C (20 mmHg)
Melting Pt: 57 °C
MDL Number: MFCD00001047
CAS-Nummer: 3032-81-3
EINECS: 221-216-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR