1-Chlor-4-fluor-2-nitrobenzol
Lieferant: ACTIVATE SCIENTIFIC
AS33781G10
AS33781G100
AS33781G5
ACTIAS33781G10EA
0
CHF
ACTIAS33781G10
ACTIAS33781G100
ACTIAS33781G5
1-Chlor-4-fluor-2-nitrobenzol
1-Chlor-4-fluor-2-nitrobenzol
Formel:
C₆H₃ClFNO₂ Siedepunkt: 238,5 °C (1013 hPa) Melting Pt: 37…40 °C |
MDL Number:
MFCD00084853 CAS-Nummer: 345-17-5 EINECS: 206-456-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...