2-Chlor-3-fluoranilin

Lieferant: ACTIVATE SCIENTIFIC

AS33743G1 AS33743G10 AS33743G5
ACTIAS33743G1EA 0 CHF
ACTIAS33743G1 ACTIAS33743G10 ACTIAS33743G5
2-Chlor-3-fluoranilin
2-Chlor-3-fluoranilin
Formel: C₆H₅ClFN
MW: 145,56 g/mol
MDL Number: MFCD03407961
CAS-Nummer: 21397-08-0
EINECS: 244-366-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR