3-Amino-4-chlorbenzoesäure
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS34844G10
AS34844G100
ACTIAS34844G10EA
0
CHF
ACTIAS34844G10
ACTIAS34844G100
3-Amino-4-chlorbenzoesäure
3-Amino-4-chlorobenzoic acid
Formel:
C₇H₆ClNO₂ Siedepunkt: 330 °C (1013 hPa) Melting Pt: 214…215 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00007671 CAS-Nummer: 2840-28-0 EINECS: 220-635-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...