2,4-Dichlorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS20697G100 AS20697G25
ACTIAS20697G100EA 0 CHF
ACTIAS20697G100 ACTIAS20697G25
2,4-Dichlorbenzonitril
2,4-Dichlorbenzonitril
Formel: C₇H₃Cl₂N
Siedepunkt: 261 °C (1013 hPa)
Melting Pt: 52…53 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00016373
CAS-Nummer: 6574-98-7
EINECS: 229-493-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR