2,3-Dichlorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS33256G10 AS33256G5
ACTIAS33256G10EA 0 CHF
ACTIAS33256G10 ACTIAS33256G5
2,3-Dichlorbenzonitril
2,3-Dichlorbenzonitril
Formel: C₇H₃Cl₂N
Siedepunkt: 70 °C (0.5 mmHg)
Melting Pt: 59 °C
MDL Number: MFCD00016372
CAS-Nummer: 6574-97-6
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR