4-Chlor-2-nitrobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS31849G1 AS31849G5
ACTIAS31849G1EA 0 CHF
ACTIAS31849G1 ACTIAS31849G5
4-Chlor-2-nitrobenzaldehyd
4-Chloro-2-nitrobenzaldehyde
Formel: C₇H₄ClNO₃
MW: 185,57 g/mol
MDL Number: MFCD00056104
CAS-Nummer: 5551-11-1
EINECS: 226-915-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR